Assay Design



Gene Transcripts

Gene Symbol
Gpr35
Entrez Gene ID
64095
Gene Name
G protein-coupled receptor 35
Gene Aliases
KPG_007
Chromosome Location
Chr.1: 92950862 - 92986391 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271766.1 NP_001258695.1 1 - 2 106 68
GenBank mRNA AK170995.1 - 1 - 2 107 68

View Full Product Details