Assay Design



Gene Transcripts

Gene Symbol
Armc9
Entrez Gene ID
78795
Gene Name
armadillo repeat containing 9
Gene Aliases
3830422A13Rik, 4831423D23Rik, 4930438O05Rik, 5730415N24Rik
Chromosome Location
Chr.1: 86154732 - 86278284 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027456.1 NP_081732.1 20 - 21 1944 60
NM_030184.2 NP_084460.1 20 - 21 1979 60
GenBank mRNA AK017564.1 - 20 - 21 1945 60
AK019600.1 - 20 - 21 1968 60
AK030307.1 - 20 - 21 1976 60
AK036083.1 - 20 - 21 1947 60
AK039800.1 - 20 - 21 1952 60
AK045699.1 - 20 - 21 1943 60
AK046693.1 - 20 - 21 1962 60
AK076239.1 - 3 - 4 210 60
AK161393.1 - 20 - 21 1959 60
AK186122.1 - 3 - 4 195 60
BC028964.1 - 13 - 14 1199 60
BC043101.1 - 20 - 21 1931 60
BQ173978.1 - 1 - 2 60

View Full Product Details