Assay Design
Gene Transcripts
- Gene Symbol
- Dedd
- Entrez Gene ID
-
21945
- Gene Name
- death effector domain-containing
- Gene Aliases
-
CASP8IP1, DEFT, Dedpro1, FLDED1, KE05
- Chromosome Location
-
Chr.1: 171328529 - 171342501 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_011615.3 |
NP_035745.3 |
1 - 2 |
377 |
68 |
View Full Product Details