Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128609.1 | NP_001122081.1 | 2 - 3 | 417 | 109 |
GenBank mRNA | AK088329.1 | - | 2 - 3 | 80 | 109 |
AK157342.1 | - | 2 - 3 | 77 | 109 | |
AK157609.1 | - | 2 - 3 | 363 | 109 | |
BM947911.1 | - | 2 - 3 | 417 | 109 |