Assay Design



Gene Transcripts

Gene Symbol
Slc41a1
Entrez Gene ID
98396
Gene Name
solute carrier family 41, member 1
Gene Aliases
AI573938, B230315F01Rik
Chromosome Location
Chr.1: 131828012 - 131848863 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173865.3 NP_776290.1 1 - 2 384 70
GenBank mRNA AK039371.1 - 1 - 2 113 70
AK045861.1 - 1 - 2 357 70
AK089802.1 - 1 - 2 385 70
AK154819.1 - 1 - 2 357 70
BC042724.1 - 1 - 2 351 70
BC096596.1 - 1 - 2 420 70
BC116280.1 - 1 - 2 192 70
BC116281.1 - 1 - 2 192 70

View Full Product Details