Assay Design



Gene Transcripts

Gene Symbol
Rcsd1
Entrez Gene ID
226594
Gene Name
RCSD domain containing 1
Gene Aliases
A430105K13Rik, BC025872
Chromosome Location
Chr.1: 165648945 - 165710063 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038846.1 NP_001033935.1 2 - 3 305 115
GenBank mRNA AK168816.1 - 2 - 3 276 115
AK171323.1 - - 285 115

View Full Product Details