Assay Design



Gene Transcripts

Gene Symbol
Slco6c1
Entrez Gene ID
74441
Gene Name
solute carrier organic anion transporter family, member 6c1
Gene Aliases
4933404A18Rik, C79179
Chromosome Location
Chr.1: 97059447 - 97128340 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028942.4 NP_083218.1 6 - 7 1230 106
GenBank mRNA AK016647.1 - 6 - 7 1178 106
AK029505.1 - 6 - 7 1231 106
AK132668.1 - 6 - 7 1181 106
AK161249.1 - 6 - 7 1181 106
BC139066.1 - 6 - 7 1133 106
BC139067.1 - 6 - 7 1133 106

View Full Product Details