Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080419.1 | NP_536344.1 | 6 - 7 | 1977 | 59 |
GenBank mRNA | AF411055.1 | - | 6 - 7 | 1848 | 59 |
AF439263.1 | - | 6 - 7 | 1960 | 59 | |
AK140254.1 | - | 6 - 7 | 1989 | 59 | |
AK146104.1 | - | 6 - 7 | 1990 | 59 | |
AK146221.1 | - | 6 - 7 | 1996 | 59 | |
AK158522.1 | - | 6 - 7 | 1998 | 59 | |
AK168363.1 | - | 6 - 7 | 1996 | 59 | |
AK189184.1 | - | 2 - 3 | 160 | 59 | |
AK190325.1 | - | 1 - 2 | 123 | 59 | |
AK216541.1 | - | 1 - 2 | 130 | 59 | |
BC019587.1 | - | 4 - 5 | 999 | 59 | |
BC026464.1 | - | 5 - 6 | 1783 | 59 | |
BC026633.1 | - | 2 - 3 | 529 | 59 | |
BC038068.1 | - | 4 - 5 | 1233 | 59 | |
BC048387.1 | - | 6 - 7 | 1977 | 59 | |
BC060087.1 | - | - | 2605 | 59 |