Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160165.1 | NP_001153637.1 | 1 - 2 | 227 | 85 |
GenBank mRNA | AB048604.1 | - | 1 - 2 | 324 | 85 |
AF139059.1 | - | 1 - 2 | 181 | 85 | |
BY088515.1 | - | 1 - 2 | 229 | 85 |