Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024495.5 | NP_078771.1 | 6 - 7 | 757 | 93 |
GenBank mRNA | AF231123.1 | - | 6 - 7 | 707 | 93 |
AK010166.1 | - | 6 - 7 | 711 | 93 | |
AK150871.1 | - | 6 - 7 | 709 | 93 | |
AK150897.1 | - | 6 - 7 | 709 | 93 | |
AK151519.1 | - | 6 - 7 | 695 | 93 | |
AK151978.1 | - | 6 - 7 | 709 | 93 | |
AK152124.1 | - | 6 - 7 | 706 | 93 | |
AK153080.1 | - | 6 - 7 | 736 | 93 | |
AK153258.1 | - | 6 - 7 | 697 | 93 | |
AK153353.1 | - | 6 - 7 | 736 | 93 | |
AK162621.1 | - | 6 - 7 | 757 | 93 | |
BC064050.1 | - | 6 - 7 | 708 | 93 |