Assay Design



Gene Transcripts

Gene Symbol
Plekhg3
Entrez Gene ID
263406
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 3
Gene Aliases
BC030417
Chromosome Location
Chr.12: 76533247 - 76579042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153804.4 NP_722499.4 18 - 19 3590 89
GenBank mRNA AK144041.1 - - 2001 89
AK145222.1 - 1 - 2 1396 89
AK172977.1 - 4 - 5 4276 89
AK192736.1 - 1 - 2 142 89
BC031136.1 - 2 - 3 1611 89
BC096443.1 - 18 - 19 3568 89

View Full Product Details