Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025840.3 | NP_080116.2 | 3 - 4 | 405 | 87 |
GenBank mRNA | AB244988.1 | - | 3 - 4 | 321 | 87 |
AK088108.1 | - | 3 - 4 | 337 | 87 | |
AK132595.1 | - | 3 - 4 | 347 | 87 | |
AK160544.1 | - | 3 - 4 | 378 | 87 | |
AK162647.1 | - | 3 - 4 | 347 | 87 | |
BC013060.1 | - | 3 - 4 | 341 | 87 | |
BC058796.1 | - | 3 - 4 | 301 | 87 |