Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009230.3 | NP_033256.2 | 2 - 3 | 301 | 95 |
GenBank mRNA | AK054149.1 | - | 2 - 3 | 214 | 95 |
AK142486.1 | - | 2 - 3 | 218 | 95 | |
AK159393.1 | - | 2 - 3 | 231 | 95 | |
AK159529.1 | - | 2 - 3 | 231 | 95 | |
AK160017.1 | - | 2 - 3 | 231 | 95 | |
AK213760.1 | - | 2 - 3 | 146 | 96 | |
BC083092.1 | - | 1 - 2 | 115 | 95 | |
CJ164331.1 | - | 2 - 3 | 304 | 95 | |
DQ903181.1 | - | 2 - 3 | 218 | 95 | |
S81092.1 | - | 2 - 3 | 235 | 95 |