Assay Design



Gene Transcripts

Gene Symbol
Cntnap1
Entrez Gene ID
53321
Gene Name
contactin associated protein-like 1
Gene Aliases
AI841080, Caspr, NCP1, Nrxn4, p190, shm
Chromosome Location
Chr.11: 101174629 - 101190727 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016782.2 NP_058062.2 17 - 18 2903 93
GenBank mRNA AF039833.1 - 17 - 18 2913 93
AK154468.1 - - 3664 93

View Full Product Details