Assay Design



Gene Transcripts

Gene Symbol
Naa15
Entrez Gene ID
74838
Gene Name
N(alpha)-acetyltransferase 15, NatA auxiliary subunit
Gene Aliases
5730450D16Rik, 6330400I15, ASTBDN, Narg1, Tbdn-1, mNAT1
Chromosome Location
Chr.3: 51416016 - 51475985 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053089.3 NP_444319.3 11 - 12 1563 134
GenBank mRNA AB221683.1 - 11 - 12 1256 134
AF237622.1 - 7 - 8 847 134
AF510858.1 - 11 - 12 1453 134
BC050017.1 - 11 - 12 1454 134

View Full Product Details