Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053089.3 | NP_444319.3 | 4 - 5 | 706 | 69 |
GenBank mRNA | AB221683.1 | - | 4 - 5 | 399 | 69 |
AF510858.1 | - | 4 - 5 | 596 | 69 | |
BC050017.1 | - | 4 - 5 | 597 | 69 | |
BQ887090.1 | - | 4 - 5 | 702 | 69 |