Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011990.2 | NP_036120.1 | 11 - 12 | 1796 | 154 |
GenBank mRNA | AB022345.1 | - | 11 - 12 | 1763 | 154 |
AK076322.1 | - | 11 - 12 | 1771 | 154 | |
AK089202.1 | - | 11 - 12 | 154 | ||
AK162233.1 | - | 11 - 12 | 1571 | 154 | |
AK163168.1 | - | 11 - 12 | 1770 | 154 | |
AK165599.1 | - | 11 - 12 | 1772 | 154 | |
AK171192.1 | - | 11 - 12 | 1794 | 154 | |
AK171252.1 | - | 11 - 12 | 1770 | 154 | |
AY766236.1 | - | 11 - 12 | 1763 | 154 | |
BC141402.1 | - | 11 - 12 | 1693 | 154 | |
BC141408.1 | - | 11 - 12 | 1693 | 154 |