Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133795.1 | NP_598556.1 | 7 - 8 | 825 | 119 |
GenBank mRNA | AK077715.1 | - | 7 - 8 | 838 | 119 |
AK166431.1 | - | 7 - 8 | 832 | 119 | |
AK170032.1 | - | 7 - 8 | 836 | 119 | |
AK170230.1 | - | 7 - 8 | 835 | 119 | |
BC010236.1 | - | 7 - 8 | 825 | 119 |