Assay Design



Gene Transcripts

Gene Symbol
Trim55
Entrez Gene ID
381485
Gene Name
tripartite motif-containing 55
Gene Aliases
D830041C10Rik, Murf2, Rnf29
Chromosome Location
Chr.3: 19644328 - 19692348 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081281.1 NP_001074750.1 5 - 6 988 64

View Full Product Details