Assay Design



Gene Transcripts

Gene Symbol
Bcap29
Entrez Gene ID
12033
Gene Name
B cell receptor associated protein 29
Gene Aliases
AW208404, Bap29
Chromosome Location
Chr.12: 31595354 - 31634658 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164090.1 NP_001157562.1 6 - 7 663 121
NM_007530.3 NP_031556.1 5 - 6 826 121
GenBank mRNA AK090068.1 - 6 - 7 640 121
AK144754.1 - 6 - 7 637 121
BC003303.1 - 5 - 6 730 121
BC021661.1 - 5 - 6 837 121
X78684.1 - 5 - 6 638 121

View Full Product Details