Assay Design



Gene Transcripts

Gene Symbol
Gabre
Entrez Gene ID
14404
Gene Name
gamma-aminobutyric acid (GABA) A receptor, subunit epsilon
Gene Aliases
-
Chromosome Location
Chr.X: 72256926 - 72274822 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017369.2 NP_059065.2 7 - 8 2188 77
GenBank mRNA AF090376.1 - 3 - 4 309 77
AF189263.1 - 7 - 8 2170 77

View Full Product Details