Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024478.1 | NP_001019649.1 | 10 - 11 | 1435 | 128 |
GenBank mRNA | AK047666.1 | - | 10 - 11 | 1436 | 128 |
AK049806.1 | - | 10 - 11 | 1437 | 128 | |
BC138911.1 | - | 10 - 11 | 1363 | 128 |