Assay Design



Gene Transcripts

Gene Symbol
Alppl2
Entrez Gene ID
11650
Gene Name
alkaline phosphatase, placental-like 2
Gene Aliases
Akp5, C77216, D1Ertd816e, EAP
Chromosome Location
Chr.1: 87086690 - 87096250 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007433.3 NP_031459.3 3 - 4 459 71
GenBank mRNA AK145571.1 - 4 - 5 368 71
AY054302.1 - 3 - 4 300 71
BC052874.1 - 3 - 4 454 71

View Full Product Details