Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029068.4 | NP_083344.3 | 7 - 8 | 1254 | 75 |
GenBank mRNA | AK015872.1 | - | 7 - 8 | 1215 | 75 |
AK032055.1 | - | 7 - 8 | 1211 | 75 | |
BC019424.1 | - | 6 - 7 | 1019 | 75 | |
BC070431.1 | - | - | 1573 | 75 |