Assay Design



Gene Transcripts

Gene Symbol
Aim2
Entrez Gene ID
383619
Gene Name
absent in melanoma 2
Gene Aliases
Gm1313, Ifi210
Chromosome Location
Chr.1: 173349246 - 173466040 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013779.2 NP_001013801.2 5 - 6 715 131
GenBank mRNA DT921984.1 - 1 - 2 241 131

View Full Product Details