Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009190.2 | NP_033216.2 | 2 - 3 | 416 | 72 |
GenBank mRNA | AK151023.1 | - | 2 - 3 | 370 | 72 |
AK152131.1 | - | 2 - 3 | 403 | 72 | |
AK165594.1 | - | - | 361 | 72 | |
AK168270.1 | - | 2 - 3 | 307 | 72 | |
BC003799.1 | - | 2 - 3 | 323 | 72 | |
U10119.1 | - | 2 - 3 | 367 | 72 |