Assay Design



Gene Transcripts

Gene Symbol
Kdsr
Entrez Gene ID
70750
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
6330410P18Rik, 9430079B08Rik, Fvt1
Chromosome Location
Chr.1: 106720410 - 106759830 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027534.2 NP_081810.1 1 - 2 198 96
GenBank mRNA AK170064.1 - 1 - 2 81 96
AY634684.1 - 1 - 2 107 96
BC023820.1 - 1 - 2 188 96

View Full Product Details