Assay Design



Gene Transcripts

Gene Symbol
Phlpp1
Entrez Gene ID
98432
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases
AI836256, Phlpp, Plekhe1, SCOP, mKIAA0606
Chromosome Location
Chr.1: 106171869 - 106394245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133821.3 NP_598582.3 1 - 2 1582 96
GenBank mRNA AB093251.1 - 1 - 2 341 96
BC059254.1 - 1 - 2 243 96
CN525246.1 - 1 - 2 48 96

View Full Product Details