Assay Design
Gene Transcripts
- Gene Symbol
- Tns4
- Entrez Gene ID
-
217169
- Gene Name
- tensin 4
- Gene Aliases
-
9930017A07Rik, AA589547, AU016405, Cten
- Chromosome Location
-
Chr.11: 99065678 - 99089306 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_172564.3 |
NP_766152.2 |
3 - 4 |
966 |
70 |
GenBank mRNA |
BC055820.1 |
- |
3 - 4 |
948 |
70 |
View Full Product Details