Assay Design
Gene Transcripts
- Gene Symbol
- Cdk5r1
- Entrez Gene ID
-
12569
- Gene Name
- cyclin-dependent kinase 5, regulatory subunit 1 (p35)
- Gene Aliases
-
Cdk5r, D11Bwg0379e, p25, p35
- Chromosome Location
-
Chr.11: 80477046 - 80481179 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK169667.1 |
- |
1 - 1 |
4082 |
95 |
BC046823.1 |
- |
1 - 1 |
4066 |
95 |
View Full Product Details