Assay Design
Gene Transcripts
- Gene Symbol
- Spata20
- Entrez Gene ID
-
217116
- Gene Name
- spermatogenesis associated 20
- Gene Aliases
-
Tisp78
- Chromosome Location
-
Chr.11: 94478904 - 94485310 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144827.4 |
NP_659076.2 |
4 - 5 |
504 |
64 |
GenBank mRNA |
BC050788.1 |
- |
4 - 5 |
504 |
64 |
View Full Product Details