Assay Design



Gene Transcripts

Gene Symbol
Lrrc59
Entrez Gene ID
98238
Gene Name
leucine rich repeat containing 59
Gene Aliases
AA959742, C78668
Chromosome Location
Chr.11: 94629824 - 94645216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133807.1 NP_598568.1 4 - 5 616 95
GenBank mRNA AK053450.1 - 4 - 5 643 95
AK159374.1 - 4 - 5 672 95
AK159552.1 - 4 - 5 666 95
AK166576.1 - 4 - 5 665 95
AK167604.1 - 4 - 5 607 95
AK169390.1 - 4 - 5 615 95
BC006877.1 - 4 - 5 616 95
BC066172.1 - 4 - 5 610 95

View Full Product Details