Assay Design



Gene Transcripts

Gene Symbol
Ovgp1
Entrez Gene ID
12659
Gene Name
oviductal glycoprotein 1
Gene Aliases
120kDa, AU016433, AU019448, Chit5, MOGP, OGP, muc9
Chromosome Location
Chr.3: 105973802 - 105987423 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007696.2 NP_031722.1 10 - 11 1172 103
GenBank mRNA AY521455.1 - 10 - 11 1158 103
BC137995.1 - 10 - 11 1263 103
D32137.1 - 10 - 11 1172 103

View Full Product Details