Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201360.1 | NP_958748.1 | 2 - 3 | 414 | 74 |
GenBank mRNA | AK078880.1 | - | 2 - 3 | 414 | 74 |
AK165474.1 | - | - | 1288 | 74 | |
BC026417.1 | - | 3 - 4 | 312 | 74 | |
BC139169.1 | - | 2 - 3 | 398 | 74 | |
BC139170.1 | - | 2 - 3 | 398 | 74 |