Assay Design



Gene Transcripts

Gene Symbol
Ccl28
Entrez Gene ID
56838
Gene Name
chemokine (C-C motif) ligand 28
Gene Aliases
CCK1, MEC, Scya28
Chromosome Location
Chr.13: 119610551 - 119654359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020279.3 NP_064675.1 1 - 2 121 91
GenBank mRNA AF220238.1 - 1 - 2 67 91
BC040510.1 - 1 - 2 133 91
BC055864.1 - 1 - 2 124 91

View Full Product Details