Assay Design
Gene Transcripts
- Gene Symbol
- Tbc1d22b
- Entrez Gene ID
-
381085
- Gene Name
- TBC1 domain family, member 22B
- Gene Aliases
-
BC045600
- Chromosome Location
-
Chr.17: 29549788 - 29606894 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198647.1 |
NP_941049.1 |
3 - 4 |
558 |
80 |
GenBank mRNA |
BC045600.1 |
- |
3 - 4 |
558 |
80 |
View Full Product Details