Assay Design



Gene Transcripts

Gene Symbol
Smc1b
Entrez Gene ID
140557
Gene Name
structural maintenance of chromosomes 1B
Gene Aliases
SMC-1B, SMC1beta, Smc1l2
Chromosome Location
Chr.15: 85064689 - 85131957 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080470.1 NP_536718.1 5 - 6 879 78
GenBank mRNA AF303827.1 - 5 - 6 879 78
BC129800.1 - 5 - 6 870 78

View Full Product Details