Assay Design



Gene Transcripts

Gene Symbol
Adamts6
Entrez Gene ID
108154
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 6
Gene Aliases
5031426K13, A930019D11Rik, ADAM-TS6, b2b1879.1Clo, b2b2029Clo, b2b2182Clo, b2b2187.1Clo, b2b2228Clo
Chromosome Location
Chr.13: 104287347 - 104496694 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081020.1 NP_001074489.1 11 - 12 1918 83
GenBank mRNA AK030315.1 - 12 - 13 2208 83
AK044528.1 - 12 - 13 2058 83
BC172116.1 - 12 - 13 1958 83

View Full Product Details