Assay Design



Gene Transcripts

Gene Symbol
Adamts6
Entrez Gene ID
108154
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 6
Gene Aliases
5031426K13, A930019D11Rik, ADAM-TS6, b2b1879.1Clo, b2b2029Clo, b2b2182Clo, b2b2187.1Clo, b2b2228Clo
Chromosome Location
Chr.13: 104287347 - 104496694 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081020.1 NP_001074489.1 14 - 15 2238 81
GenBank mRNA AK044528.1 - 15 - 16 2378 81

View Full Product Details