Assay Design



Gene Transcripts

Gene Symbol
Sycp1
Entrez Gene ID
20957
Gene Name
synaptonemal complex protein 1
Gene Aliases
SCP1
Chromosome Location
Chr.3: 102818499 - 102936312 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011516.2 NP_035646.2 5 - 6 413 78
GenBank mRNA AK133229.1 - 5 - 6 414 78
BC137967.1 - 5 - 6 375 78
L41069.1 - 5 - 6 394 78
Z38118.1 - 5 - 6 404 78

View Full Product Details