Assay Design



Gene Transcripts

Gene Symbol
Nnt
Entrez Gene ID
18115
Gene Name
nicotinamide nucleotide transhydrogenase
Gene Aliases
4930423F13Rik, AI323702, BB168308
Chromosome Location
Chr.13: 119335527 - 119409138 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008710.3 NP_032736.2 - 317 75
GenBank mRNA AK033282.1 - 2 - 3 258 75
AK040460.1 - 2 - 3 278 75
AK050478.1 - 1 - 2 222 75
AK052061.1 - 2 - 3 256 75
AK052234.1 - 2 - 3 302 75
AK054332.1 - 2 - 3 303 75
AK083413.1 - 2 - 3 303 75
AK084546.1 - 2 - 3 300 75
AK084604.1 - 2 - 3 300 75
AK085903.1 - 2 - 3 303 75
AK087064.1 - 2 - 3 299 75
AK088044.1 - 2 - 3 299 75
AK090115.1 - - 320 75
AK129006.1 - 2 - 3 294 75
AK159694.1 - 2 - 3 303 75
BC008518.1 - 2 - 3 280 75
Z49204.1 - 1 - 2 224 75

View Full Product Details