Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289442.1 | NP_001276371.1 | 3 - 4 | 325 | 74 |
NM_010815.3 | NP_034945.1 | 2 - 3 | 420 | 74 | |
GenBank mRNA | AF053405.1 | - | 2 - 3 | 388 | 74 |
AF055465.1 | - | 2 - 3 | 420 | 74 | |
AF129477.1 | - | 2 - 3 | 383 | 74 | |
AF236118.1 | - | 2 - 3 | 390 | 74 | |
AJ011735.1 | - | 2 - 3 | 386 | 74 | |
AK156939.1 | - | 2 - 3 | 399 | 74 | |
BC034859.1 | - | 3 - 4 | 562 | 74 | |
BC052496.1 | - | 3 - 4 | 337 | 74 |