Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080769.1 | NP_001074238.1 | 17 - 18 | 3939 | 68 |
GenBank mRNA | AK089790.1 | - | 17 - 18 | 3967 | 68 |
AK144200.1 | - | 12 - 13 | 3235 | 68 | |
AK220293.1 | - | 17 - 18 | 3939 | 68 |