Assay Design



Gene Transcripts

Gene Symbol
Slc26a8
Entrez Gene ID
224661
Gene Name
solute carrier family 26, member 8
Gene Aliases
-
Chromosome Location
Chr.17: 28637779 - 28691237 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290320.1 NP_001277249.1 11 - 12 1642 81
GenBank mRNA BC027076.1 - 11 - 12 1642 81

View Full Product Details