Assay Design



Gene Transcripts

Gene Symbol
Scube3
Entrez Gene ID
268935
Gene Name
signal peptide, CUB domain, EGF-like 3
Gene Aliases
CEGF3, D030038I21Rik
Chromosome Location
Chr.17: 28142100 - 28174851 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004366.1 NP_001004366.1 6 - 7 710 77
GenBank mRNA AY639609.1 - 6 - 7 710 77
BC079849.1 - 4 - 5 459 77

View Full Product Details