Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172592.2 | NP_766180.1 | 1 - 2 | 695 | 78 |
GenBank mRNA | AK033354.1 | - | 1 - 2 | 695 | 78 |
AK037115.1 | - | 1 - 2 | 671 | 78 | |
AK039311.1 | - | 3 - 4 | 430 | 78 | |
AK052265.1 | - | 3 - 4 | 564 | 78 | |
AK054414.1 | - | 1 - 2 | 1146 | 78 | |
BC063761.1 | - | 4 - 5 | 440 | 78 | |
BC070460.1 | - | 1 - 2 | 68 | 78 |