Assay Design



Gene Transcripts

Gene Symbol
Tbx21
Entrez Gene ID
57765
Gene Name
T-box 21
Gene Aliases
TBT1, Tbet, Tblym
Chromosome Location
Chr.11: 97098007 - 97115331 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019507.2 NP_062380.2 2 - 3 836 80
GenBank mRNA AF093099.1 - 2 - 3 794 80
AF241242.1 - 2 - 3 826 80
AK054495.1 - 2 - 3 835 80
AK156271.1 - 2 - 3 837 80
AY836551.1 - 1 - 2 149 80
BC137986.1 - 2 - 3 754 80
BC137988.1 - 2 - 3 754 80

View Full Product Details