Assay Design



Gene Transcripts

Gene Symbol
Tbx21
Entrez Gene ID
57765
Gene Name
T-box 21
Gene Aliases
TBT1, Tbet, Tblym
Chromosome Location
Chr.11: 97098007 - 97115331 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019507.2 NP_062380.2 3 - 4 958 116
GenBank mRNA AF093099.1 - 3 - 4 916 116
AF241242.1 - 3 - 4 948 116
AK054495.1 - 3 - 4 957 116
AK156271.1 - 3 - 4 959 116
BC137986.1 - 3 - 4 876 116
BC137988.1 - 3 - 4 876 116

View Full Product Details