Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205345.1 | NP_001192274.1 | 12 - 13 | 1635 | 79 |
NM_001205346.1 | NP_001192275.1 | 12 - 13 | 1629 | 79 | |
NM_194342.3 | NP_919323.2 | 11 - 12 | 1539 | 79 | |
GenBank mRNA | AK128958.1 | - | 11 - 12 | 1533 | 79 |
AK156246.1 | - | 12 - 13 | 1636 | 79 | |
AK171058.1 | - | 12 - 13 | 1598 | 79 | |
AK172398.1 | - | 12 - 13 | 1637 | 79 | |
AY682987.1 | - | 11 - 12 | 1395 | 79 | |
BC023965.1 | - | 1 - 2 | 187 | 79 |