Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205345.1 | NP_001192274.1 | 17 - 18 | 2315 | 79 |
NM_001205346.1 | NP_001192275.1 | 17 - 18 | 2309 | 79 | |
NM_194342.3 | NP_919323.2 | 16 - 17 | 2219 | 79 | |
GenBank mRNA | AK047688.1 | - | 5 - 6 | 661 | 79 |
AK128958.1 | - | 16 - 17 | 2213 | 79 | |
AK156246.1 | - | 17 - 18 | 2316 | 79 | |
AK171058.1 | - | 17 - 18 | 2278 | 79 | |
AK172398.1 | - | 17 - 18 | 2317 | 79 | |
AK172996.1 | - | 15 - 16 | 2919 | 79 | |
AY682987.1 | - | 16 - 17 | 2075 | 79 | |
BC023965.1 | - | 6 - 7 | 867 | 79 | |
BC098208.1 | - | 16 - 17 | 2560 | 79 | |
BQ879674.1 | - | 4 - 5 | 431 | 79 |